Recombinant isocitrate dehydrogenase
Lieferant: Apollo Scientific
BITP1329-1MG BITP1329-50MG BITP1329-5MG
APOSBITP1329-1MGEA 0 EUR
APOSBITP1329-1MG APOSBITP1329-50MG APOSBITP1329-5MG
Recombinant isocitrate dehydrogenase
Enzyme
For long term storage it is recommended to add a carrier protein (0.1% HAS or BSA). Please avoid freeze-thaw cycles.