Recombinant isocitrate dehydrogenase
Lieferant: Apollo Scientific
BITP1329-1MG
BITP1329-50MG
BITP1329-5MG
APOSBITP1329-1MGEA
0
EUR
APOSBITP1329-1MG
APOSBITP1329-50MG
APOSBITP1329-5MG
Recombinant isocitrate dehydrogenase
Enzyme
For long term storage it is recommended to add a carrier protein (0.1% HAS or BSA). Please avoid freeze-thaw cycles.