Natriumethanolat 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Ethanol Natriumsalz, Natriumethylat, Natriumethoxid

IN1022-500G
APOSIN1022-500GEA 120 EUR
APOSIN1022-500G
Natriumethanolat 96%
Natriumethanolat
Formel: C₂H₅NaO
Molecular Weight: 68,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012417
CAS-Nummer: 141-52-6
UN: 3206
ADR: 4.2+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR