Aluminiumnitrat Hydrat 99.999%
Lieferant: Apollo Scientific
|
Achtung
|
IN1027-100G
IN1027-500G
APOSIN1027-100GEA
0
EUR
APOSIN1027-100G
APOSIN1027-500G
Aluminiumnitrat Hydrat 99.999%
Aluminiumnitrat Hydrat
Formel:
AlN₃O₉·H₂O Siedepunkt: 130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003419 CAS-Nummer: 25838-59-9 UN: 1438 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...