Ammonium tetrathiomolybdate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ammonium tetrathiomolybdate(VI)

IN1087-10G IN1087-50G IN1087-1G
APOSIN1087-10GEA 306 EUR
APOSIN1087-10G APOSIN1087-50G APOSIN1087-1G
Ammonium tetrathiomolybdate
Ammonium tetrathiomolybdate
Formel: H₁₀MoN₂S₄
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00136013
CAS-Nummer: 15060-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR