Ammoniumthiocyanat 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ammoniumsulfocyanid, Ammoniumrhodanid
IN1093-1KG
APOSIN1093-1KGEA
0
EUR
APOSIN1093-1KG
Ammoniumthiocyanat 98+%
Ammoniumthiocyanat
Formel:
NH₄SCN Molecular Weight: 76,12 g/mol Schmelzpunkt: 152…154 °C Dichte: 1,3 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011428 CAS-Nummer: 1762-95-4 EINECS: 217-175-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...