Bariumwolframat 99,9%
Lieferant: Apollo Scientific
IN1228-100G
IN1228-50G
APOSIN1228-100GEA
493
EUR
APOSIN1228-100G
APOSIN1228-50G
Bariumwolframat 99,9%
Bariumwolframat
Formel:
BaO₄W Dichte: 5,04 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
CAS-Nummer:
7787-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...