Boron phosphate 99,9%

Lieferant: Apollo Scientific
Achtung

IN1296-100G
APOSIN1296-100GEA 319 EUR
APOSIN1296-100G
Boron phosphate 99,9%
Boron phosphate
Formel: BO₄P
Molecular Weight: 105,78 g/mol
Schmelzpunkt: >1400 °C
Dichte: 2,81 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011318
CAS-Nummer: 13308-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR