Certrichlorid Hydrat 99,9%
Lieferant: Apollo Scientific
|
Achtung
|
IN1414-250G
APOSIN1414-250GEA
0
EUR
APOSIN1414-250G
Certrichlorid Hydrat 99,9%
Certrichlorid Hydrat
Formel:
CeCl₃·xH₂O Schmelzpunkt: 848 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149633 CAS-Nummer: 19423-76-8 EINECS: 232-227-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...