Chromium(II) acetate hydrate

Lieferant: Apollo Scientific
Achtung

IN1462-25G
APOSIN1462-25GEA 290 EUR
APOSIN1462-25G
Chromium(II) acetate hydrate
Formel: C₄H₆CrO₄·H₂O
Dichte: 1,79 g/cm³ (25 °C)
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150821
CAS-Nummer: 628-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR