Kupfer(I)bromid-Dimethylsulfid-Komplex 99%

Lieferant: Apollo Scientific
Achtung

IN1541-10G
APOSIN1541-10GEA 0 EUR
APOSIN1541-10G
Kupfer(I)bromid-Dimethylsulfid-Komplex 99%
Kupfer(I)bromid-Dimethylsulfid-Komplex
Formel: C₂H₆BrCuS
Lagertemperatur: Raumtemperatur
MDL: MFCD00043295
CAS-Nummer: 54678-23-8
UN: 3335
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR