Copper(II) ethoxide
Lieferant: Apollo Scientific
IN1550-10G
APOSIN1550-10GEA
142
EUR
APOSIN1550-10G
Copper(II) ethoxide
Copper(II) ethoxide
Formel:
C₄H₁₀CuO₂ Molecular Weight: 153,67 g/mol Schmelzpunkt: 120 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015650 CAS-Nummer: 2850-65-9 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...