Dysprosiumtrichlorid Hexahydrat 99,9%

Lieferant: Apollo Scientific
Achtung

IN1639-100G IN1639-25G IN1639-250G
APOSIN1639-100GEA 0 EUR
APOSIN1639-100G APOSIN1639-25G APOSIN1639-250G
Dysprosiumtrichlorid Hexahydrat 99,9%
Dysprosiumtrichlorid Hexahydrat
Formel: Cl₃Dy·6H₂O
Schmelzpunkt: 718 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010984
CAS-Nummer: 15059-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR