Dysprosiumnitrid 99%

Lieferant: Apollo Scientific

IN1654-1G
APOSIN1654-1GEA 79.9 EUR
APOSIN1654-1G
Dysprosiumnitrid 99%
Dysprosium nitride
Formel: DyN
Lagertemperatur: Raumtemperatur
CAS-Nummer: 12019-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR