Dysprosium(III) oxalate hydrate 99.99+%

Lieferant: Apollo Scientific

IN1657-10G
APOSIN1657-10GEA 94.4 EUR
APOSIN1657-10G
Dysprosium(III) oxalate hydrate 99.99+%
Dysprosium(III) oxalate hydrate
Formel: C₆Dy₂O₁₂·H₂O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 58176-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR