Erbium(III) acetate hydrate 99,99%
Lieferant: Apollo Scientific
IN1684-50G
APOSIN1684-50GEA
284
EUR
APOSIN1684-50G
Erbium(III) acetate hydrate 99,99%
Erbium(III) acetate hydrate
Formel:
(C₂H₃O₂)₃·Er·H₂O Dichte: 2,114 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150113 CAS-Nummer: 207234-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...