Erbiumtrichlorid Hexahydrat 99.99%

Lieferant: Apollo Scientific
Achtung

IN1696-100G IN1696-25G
APOSIN1696-100GEA 270 EUR
APOSIN1696-100G APOSIN1696-25G
Erbiumtrichlorid Hexahydrat 99.99%
Erbiumtrichlorid Hexahydrat
Formel: Cl₃Er·6H₂O
Molecular Weight: 381.71 g/mol
Schmelzpunkt: 774 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149693
CAS-Nummer: 10025-75-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR