Erbium(III) oxalate hydrate 99,9%

Lieferant: Apollo Scientific

IN1723-25G
APOSIN1723-25GEA 219 EUR
APOSIN1723-25G
Erbium(III) oxalate hydrate 99,9%
Erbium(III) oxalate hydrate
Formel: (C₂O₄)₃·Er₂·H₂O
Molecular Weight: 616,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150141
CAS-Nummer: 58176-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR