Erbium(III)oxalat Decahydrat 99.99%
Lieferant: Apollo Scientific
IN1726-25G
APOSIN1726-25GEA
109
EUR
APOSIN1726-25G
Erbium(III)oxalat Decahydrat 99.99%
Erbium(III)oxalat Decahydrat
Formel:
C₆Er₂O₁₂·10H₂O Molecular Weight: 778,73 g/mol (598,58 anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150142 CAS-Nummer: 30618-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...