Erbium(III) phosphate 99%

Lieferant: Apollo Scientific

IN1735-1G
APOSIN1735-1GEA 50.9 EUR
APOSIN1735-1G
Erbium(III) phosphate 99%
Erbium(III) phosphate
Formel: ErPO₄
Molecular Weight: 262,23 g/mol
Schmelzpunkt: 120 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00215989
CAS-Nummer: 14298-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR