Europiumtrinitrat Hydrat 99.99%
Lieferant: Apollo Scientific
|
Achtung
|
IN1780-25G
IN1780-5G
IN1780-100G
APOSIN1780-25GEA
0
EUR
APOSIN1780-25G
APOSIN1780-5G
APOSIN1780-100G
Europiumtrinitrat Hydrat 99.99%
Europiumtrinitrat Hydrat
Formel:
EuN₃O₉·xH₂O Schmelzpunkt: 85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149697 CAS-Nummer: 100587-95-9 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...