Gadolinium ≥99,99%

Lieferant: Apollo Scientific

IN1801-1KG
APOSIN1801-1KGEA 6440 EUR
APOSIN1801-1KG
Gadolinium ≥99,99%
Gadolinium
Formel: Gd
Molecular Weight: 157,25 g/mol
Siedepunkt: 3233 °C
Schmelzpunkt: 1312 °C
MDL: MFCD00011022
CAS-Nummer: 7440-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR