Gadolinium(III)chlorid Hexahydrat 99,9%

Lieferant: Apollo Scientific
Achtung

IN1822-100G IN1822-5G
APOSIN1822-100GEA 0 EUR
APOSIN1822-100G APOSIN1822-5G
Gadolinium(III)chlorid Hexahydrat 99,9%
Gadolinium(III)chlorid Hexahydrat
Formel: Cl₃Gd·6H₂O
Dichte: 2,424 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149732
CAS-Nummer: 13450-84-5
EINECS: 233-386-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR