Gadolinium(III)-2,4-pentanedionate hydrate 99,9%
Lieferant: Apollo Scientific
|
Achtung
|
IN1840-5G
APOSIN1840-5GEA
125
EUR
APOSIN1840-5G
Gadolinium(III)-2,4-pentanedionate hydrate 99,9%
Gadolinium(III)-2,4-pentanedionate hydrate
Formel:
C₁₅H₂₁GdO₆·xH₂O Molecular Weight: 454,58 g/mol (anhy) Schmelzpunkt: 143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013493 CAS-Nummer: 64438-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...