Gallium(III)bromid, wasserfrei 99,9%
Lieferant: Apollo Scientific
Synonyme:
Gallium (III) bromide
IN1864-25G
APOSIN1864-25GEA
796
EUR
APOSIN1864-25G
Gallium(III)bromid, wasserfrei 99,9%
Gallium(III)bromid
Formel:
Br₃Ga Molecular Weight: 309,44 g/mol Siedepunkt: 279 °C Schmelzpunkt: 121,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016100 CAS-Nummer: 13450-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...