Holmium 99,9%, Pulver –40 mesh
Lieferant: Apollo Scientific
IN1966-25G
IN1966-5G
APOSIN1966-25GEA
1450
EUR
APOSIN1966-25G
APOSIN1966-5G
Holmium 99,9%, Pulver –40 mesh
Holmium
Formel:
Ho Molecular Weight: 164.93 g/mol Siedepunkt: 2600 °C Schmelzpunkt: 1474 °C Dichte: 8.8 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011049 CAS-Nummer: 7440-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...