Iron(III) oxalate hexahydrate 99.99%
Lieferant: Apollo Scientific
|
Achtung
|
IN2080-100G
APOSIN2080-100GEA
143
EUR
APOSIN2080-100G
Iron(III) oxalate hexahydrate 99.99%
Iron(III) oxalate hexahydrate
Formel:
Fe₂(C₂O₄)₃·6H₂O Schmelzpunkt: 100 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054430 CAS-Nummer: 166897-40-1 EINECS: 220-951-7 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...