Lithiumthiocyanat 99,9%

Lieferant: Apollo Scientific
Achtung

Synonyme: Lithiumrhodanid, Lithiumsulfocyanid

IN2380-25G
APOSIN2380-25GEA 139 EUR
APOSIN2380-25G
Lithiumthiocyanat 99,9%
Lithiumthiocyanat
Formel: CLiNS
Molecular Weight: 65,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011097
CAS-Nummer: 556-65-0
EINECS: 209-135-1
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR