Magnesium formate dihydrate 98+%

Lieferant: Apollo Scientific
Achtung

IN2449-100G
APOSIN2449-100GEA 125 EUR
APOSIN2449-100G
Magnesium formate dihydrate 98+%
Magnesium formate dihydrate
Formel: C₂H₂MgO₄·2H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150512
CAS-Nummer: 6150-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR