Potassium hexacyanoruthenate(II) trihydrate 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium hexacyanoruthenate trihydrate

IN2911-1G
APOSIN2911-1GEA 219 EUR
APOSIN2911-1G
Potassium hexacyanoruthenate(II) trihydrate 99%
Potassium hexacyanoruthenate(II) trihydrate
Formel: C₆K₄N₆Ru·3H₂O
Molecular Weight: 467,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00168064
CAS-Nummer: 15002-31-0
UN: 1588
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR