Praseodymtrinitrat Hydrat 99,9%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Praseodym(III)nitrat

IN2974-25G
APOSIN2974-25GEA 277 EUR
APOSIN2974-25G
Praseodymtrinitrat Hydrat 99,9%
Praseodymtrinitrat Hydrat
Formel: Pr(NO₃)₃·xH₂O
Molecular Weight: 326.92 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011175
CAS-Nummer: 15878-77-0
UN: 1477
ADR: 5.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR