Dipraseodymtrisulfat Octahydrat 99,9%

Lieferant: Apollo Scientific

IN2986-25G
APOSIN2986-25GEA 98.6 EUR
APOSIN2986-25G
Dipraseodymtrisulfat Octahydrat 99,9%
Dipraseodymtrisulfat Octahydrat
Formel: O₁₂Pr₂S₃·8H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149832
CAS-Nummer: 13510-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR