Rhodium(II)acetat dimer Rh 46,6%
Lieferant: Apollo Scientific
|
Achtung
|
IN3003-250MG
APOSIN3003-250MGEA
0
EUR
APOSIN3003-250MG
Rhodium(II)acetat dimer Rh 46,6%
Rhodium(II)acetat dimer
Formel:
C₈H₁₂O₈Rh₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003538 CAS-Nummer: 15956-28-2 EINECS: 240-084-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...