Dichloro-(p-cymol)-ruthenium(II) Dimer 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)

IN3047-1G
APOSIN3047-1GEA 40.6 EUR
APOSIN3047-1G
Dichloro-(p-cymol)-ruthenium(II) Dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer
Formel: C₂₀H₂₈Cl₄Ru₂
Molecular Weight: 612,38 g/mol
MDL: MFCD00064793
CAS-Nummer: 52462-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR