Scandium(III) acetate hydrate 99.99%

Lieferant: Apollo Scientific

Synonyme: Acetic acid scandium(III) salt hydrate

IN3118-1G
APOSIN3118-1GEA 255 EUR
APOSIN3118-1G
Scandium(III) acetate hydrate 99.99%
Scandium(III) acetate hydrate
Formel: C₆H₉O₆Sc·H₂O
Molecular Weight: 240.10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150122
CAS-Nummer: 304675-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR