Terbium(III)fluorid 99,9%

Lieferant: Apollo Scientific

Synonyme: Terbiumtrifluorid

IN3478-25G IN3478-5G
APOSIN3478-25GEA 343 EUR
APOSIN3478-25G APOSIN3478-5G
Terbium(III)fluorid 99,9%
Terbium(III)fluorid
Formel: TbF₃
Schmelzpunkt: 1172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049572
CAS-Nummer: 13708-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR