Terbium(III) oxalate decahydrate 99,9%

Lieferant: Apollo Scientific
Achtung

Synonyme: Terbium(III) ethane-1,2-dioate decahydrate

IN3484-10G
APOSIN3484-10GEA 149 EUR
APOSIN3484-10G
Terbium(III) oxalate decahydrate 99,9%
Terbium(III) oxalate decahydrate
Formel: 3(C₂O₄)·Tb₂·10H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00748140
CAS-Nummer: 24670-06-2
UN: 3288
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR