Terbium(III,IV)oxid 99,9%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tetraterbium heptaoxide, Terbium oxide
IN3487-25G
IN3487-5G
IN3487-100G
APOSIN3487-25GEA
261
EUR
APOSIN3487-25G
APOSIN3487-5G
APOSIN3487-100G
Terbium(III,IV)oxid 99,9%
Terbium(III,IV)oxid
Formel:
O₇Tb₄ Schmelzpunkt: 2340 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00083151 CAS-Nummer: 12037-01-3 EINECS: 234-856-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...