EINECS 238-195-1 99,9%
Lieferant: Apollo Scientific
Synonyme:
Terbium(III) 2,4-pentanedionate
IN3490-2G
APOSIN3490-2GEA
226
EUR
APOSIN3490-2G
EINECS 238-195-1 99,9%
Terbium(III) acetylacetonate
Formel:
C₁₅H₂₁O₆Tb Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013503 CAS-Nummer: 14284-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...