Terbium(III)sulfat Octahydrat 99,9%

Lieferant: Apollo Scientific

Synonyme: Diterbiumtrisulfat Octahydrat

IN3493-10G
APOSIN3493-10GEA 306 EUR
APOSIN3493-10G
Terbium(III)sulfat Octahydrat 99,9%
Terbium(III)sulfat Octahydrat
Formel: O₁₄S₃Tb₂·8H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149872
CAS-Nummer: 13842-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR