Dithuliumtricarbonat Hydrat 99,9%
Lieferant: Apollo Scientific
|
Achtung
|
IN3595-2G
APOSIN3595-2GEA
143
EUR
APOSIN3595-2G
Dithuliumtricarbonat Hydrat 99,9%
Dithuliumtricarbonat Hydrat
Formel:
C₃O₉Tm₂·H₂O Lagertemperatur: Raumtemperatur |
CAS-Nummer:
87198-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...