Thulium(III)chlorid Hydrat 99,9%

Lieferant: Apollo Scientific
Achtung

IN3598-5G
APOSIN3598-5GEA 190 EUR
APOSIN3598-5G
Thulium(III)chlorid Hydrat 99,9%
Thulium(III)chlorid Hydrat
Formel: Cl₃Tm·H₂O
Molecular Weight: 293.309 g/mol
Schmelzpunkt: 824 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149877
CAS-Nummer: 19423-86-0
EINECS: 236-904-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR