Thulium(III) oxalate hydrate 99,9%

Lieferant: Apollo Scientific
Achtung

IN3610-2G
APOSIN3610-2GEA 174 EUR
APOSIN3610-2G
Thulium(III) oxalate hydrate 99,9%
Thulium(III) oxalate hydrate
Formel: C₆O₁₂Tm₂·xH₂O
Molecular Weight: 601,93 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00050804
CAS-Nummer: 58176-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR