Thulium(III)oxid 99.99%

Lieferant: Apollo Scientific
Achtung

IN3616-1G IN3616-5G
APOSIN3616-1GEA 125 EUR
APOSIN3616-1G APOSIN3616-5G
Thulium(III)oxid 99.99%
Thulium(III)oxid
Formel: O₃Tm₂
Schmelzpunkt: 2425 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011285
CAS-Nummer: 12036-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR