Tin(IV) chloride hydrate 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Stannic chloride, Tin tetrachloride hydrate

IN3650-25G
APOSIN3650-25GEA 46.4 EUR
APOSIN3650-25G
Tin(IV) chloride hydrate 98%
Tin(IV) chloride hydrate
Formel: SnCl₄·xH₂O
Molecular Weight: 260,50 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00216885
CAS-Nummer: 7646-78-8
UN: 2440
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR