Ytterbiumtriacetat Tetrahydrat 99,9%

Lieferant: Apollo Scientific

IN3787-5G
APOSIN3787-5GEA 94.4 EUR
APOSIN3787-5G
Ytterbiumtriacetat Tetrahydrat 99,9%
Ytterbiumtriacetat Tetrahydrat
Formel: C₆H₉O₆Yb·4H₂O
Dichte: 2,09 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 15280-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR