Ytterbiumtrinitrat 99,9%

Lieferant: Apollo Scientific
Gefahr

IN3805-50G
APOSIN3805-50GEA 212 EUR
APOSIN3805-50G
Ytterbiumtrinitrat 99,9%
Ytterbiumtrinitrat
Formel: N₃O₉Yb
Molecular Weight: 359,05 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 13768-67-7
UN: 1477
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR