Ytterbium(III) 2,4-pentanedionate 99,9%

Lieferant: Apollo Scientific

Synonyme: Ytterbium(III) acetylacetonate

IN3814-2G
APOSIN3814-2GEA 136 EUR
APOSIN3814-2G
Ytterbium(III) 2,4-pentanedionate 99,9%
Ytterbium(III) 2,4-pentanedionate
Formel: C₁₅H₂₁O₆Yb
Molecular Weight: 470,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013508
CAS-Nummer: 14284-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR