Yttrium(III) acetate tetrahydrate 99.99%

Lieferant: Apollo Scientific
Achtung

IN3832-25G
APOSIN3832-25GEA 105 EUR
APOSIN3832-25G
Yttrium(III) acetate tetrahydrate 99.99%
Yttrium(III) acetate tetrahydrate
Formel: C₆H₉O₆Y·4H₂O
Molecular Weight: 338,10 g/mol (266,04 anhy)
Dichte: 1,5 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013048
CAS-Nummer: 85949-60-6
EINECS: 245-612-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR