Yttrium(III)nitrat Hexahydrat 99,9%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Yttriumtrinitrat Hexahydrat

IN3856-100G
APOSIN3856-100GEA 132 EUR
APOSIN3856-100G
Yttrium(III)nitrat Hexahydrat 99,9%
Yttrium(III)nitrat Hexahydrat
Formel: N₃O₉Y·6H₂O
Molecular Weight: 383,01 g/mol
Dichte: 2,682 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149940
CAS-Nummer: 13494-98-9
EINECS: 233-802-6
UN: 1477
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR